Learn More
Description
Includes
- 10x Wash Buffer, 25mL
- Blocking Buffer, 25mL
- ALP Substrate, 6mL
- HRP Substrate, 6mL
- ERK-Ab1, 10μL
- pERK-Ab1, 10μL
- HRP-Ab2, 10μL
- ALP-Ab2, 10μL
- Safe, non-radioactive assay
- Simple and convenient. Total and pERK can be measured in the same sample
Specifications
Specifications
| Cross Reactivity | Human, Mouse, Rat |
| Description | ERK Phosphorylation Cell-Based ELISA Kit is an indirect enzyme-linked immunoassay for qualitative determination of ERK phosphorylation in cultured cells. |
| Detection Method | Fluorimetric |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Sample Type | Cell |
| Content And Storage | Store the kit at -20°C |
| For Use With (Application) | Quantification |
| Product Type | ELISA Kit |
| No. of Assays | 100 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.