Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ESR1 (Human) ELISA Kit

Catalog No. 89100739 Shop All Abnova Corporation Products
Catalog No. 89-100-739 Supplier Abnova™ Supplier No. KA3032

Includes

  • One 96-well plate pre-coated with anti-human Estrogen antibody, 96 (8 x 12) wells
  • Standard (13.5ng/mL), 0.5mL
  • Standard diluent buffer, 1.5mL
  • Wash buffer (30x), Dilution: 1:30, 20mL
  • Sample diluent buffer, 6mL
  • HRP conjugated anti-human Estrogen antibody (RTU), 6mL
  • Stop solution, 6mL
  • TMB substrate A, 6mL
  • TMB substrate B, 6mL
  • Plate sealer, 2 slices
  • Hermetic bag, 1 bag

Use to measure quantitatively human ESR1.

This kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 0.3 to 10ng/mL
Conjugate HRP
Cross Reactivity Human
Description ESR1 (Human) ELISA Kit is intended for the quantitative measurement of human ESR1.
Detection Limit 10pg/mL
Detection Method Colorimetric
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Sample, Urine
Sample Volume 10 μL
Packaging 96-well
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type ESR1 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.