Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Estradiol (Canine) ELISA Kit

Catalog No. 89100548 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-548 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-548 Supplier Abnova™ Supplier No. KA2288
Add to Cart
Add to Cart

Includes

  • Secondary antibody-coated microtiter wells, 96 wells
  • HRP Conjugate, 12mL
  • TMB Color Reagent, 12mL
  • Stopping solution (2N HCl), 6mL
  • 20x Wash buffer, 20mL
  • Standard/Sample diluent, 20mL
  • Standard Set (Lyophilized) (0, 10, 25, 100, 500, 2500 and 10,000pg/mL) dissolve in onemL using standard/sample diluent, 7 vials
  • Quality control set QC1, (∼100pg/mL) and QC2, (∼500pg/mL) dissolve using 1mL standard/sample diluent, 1 vial/each

Use to measure quantitatively estradiol in canine serum, plasma. Enzyme immunoassay

The E2 quantitative test is based on a solid-phase enzyme immunoassay based on competitive binding method

Specifications

Calibration Range 0 to 10,000pg/mL
Conjugate HRP
Cross Reactivity Dog
Description Estradiol (Canine) ELISA Kit is an enzyme immunoassay for quantitative determination of estradiol in canine serum, plasma.
Detection Limit 5pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Serum
Sample Volume 50 μL
Sensitivity 5 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.