Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Estradiol (Horse) ELISA Kit

Catalog No. 89100558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-558 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-558 Supplier Abnova™ Supplier No. KA2299
Add to Cart
Add to Cart

Includes

  • Microtiter wells, coated with second antibody, 96 wells
  • Lyophilized Standard Set: 0, 10, 25, 100, 500, 2500, 10,000pg/mL (QC1 range 100pg/mL and QC2 range 500pg/mL), reconstitute in 1.0mL Standard/Sample diluent, 1 set
  • Enzyme Conjugate Solution, 12mL
  • TMB Color Reagent, 12mL
  • Stopping solution (2N HCl), 6mL
  • 20x Wash buffer, 20mL
  • Standard/Sample diluent, 20mL

Use to measure quantitatively estradiol in horse serum, plasma, urine. Enzyme immunoassay.

The Estradiol (Horse) ELISA Kit is based on a solid-phase enzyme immunoassay based on competitive binding method

Specifications

Calibration Range 10 to 10,000pg/mL
Conjugate HRP
Cross Reactivity Horse
Description Estradiol (Horse) ELISA Kit is an enzyme immunoassay for quantitative determination of estradiol in horse serum, plasma, urine.
Detection Limit 10pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 25 μL
Sensitivity 10 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.