Learn More
Description
Includes
- EV71 Ab Coated Well, 1 plate
- EV71-VPI Standard, Lyophilized, 3 vials
- Conjugate Concentrate (100x), 0.15mL
- Conjugate Diluent, 18mL
- Standard / Sample Diluent, 12mL x 3
- TMB Reagent, 11mL
- Stop Solution (1N HCl), 11mL
- Wash Buffer (20x), 50mL
Specifications
Specifications
| Calibration Range | 1 to 100ng/mL |
| Conjugate | HRP |
| Description | EV71 VP1 ELISA Kit is an Enzyme-Linked Immunosorbent Assay for the quantitative measurement of EV71 VP1. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Content And Storage | Store the kit at 2-8°C. |
| For Use With (Application) | Quantification |
| Product Type | EV71 VP1 ELISA Kit |
| No. of Assays | 96 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.