Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ F2 (Mouse) ELISA Kit

Catalog No. 89100369 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-369 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-369 Supplier Abnova™ Supplier No. KA2111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Mouse Prothrombin Microplate
  • Sealing Tapes
  • Mouse Prothrombin Standard: Mouse Prothrombin in a buffered protein base, 50ng, lyophilized
  • Biotinylated Mouse Prothrombin Antibody (50x): A 50-fold concentrated biotinylated polyclonal antibody against Mouse Prothrombin
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant, 30mL, 2 bottles
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate, 80μL
  • Chromogen Substrate
  • Stop Solution: A 0.5N hydroxychloric acid

Use to measure quantitatively mouse F2. Sandwich enzyme immunoassay.

This assay employs a quantitative sandwich enzyme immunoassay technique that measures mouse Prothrombin in less than 4 hours

Specifications

Conjugate Biotin
Cross Reactivity Mouse
Description F2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse F2.
Detection Limit 0.7ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store SP Conjugate and Biotinylated Antibody at -20°C. Store Standard at 2-8°C before reconstituting with Diluent and at -20°C after reconstituting with Diluent. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.