Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ F2 (Pig) ELISA Kit

Catalog No. 89024527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-527 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-527 Supplier Abnova™ Supplier No. KA1407
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Pig F2 in Plasma, Serum and cell culture supernatants

Specifications

Calibration Range 1 to 100ng/mL
Conjugate Biotin
Cross Reactivity Pig
Description F2 (Pig) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of pig F2 in plasma, serum and cell culture supernatants.
Detection Method Colorimetric
Includes 96 Antibody coated microwell, 800μg swine prothrombin standard, 80μL Biotinylated swine prothrombin Antibody (100X), 30mL EIA diluent (10X), 2 30mL Wash Buffer (20X), 80μL Streptavidin-Peroxidase Conju
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume 50 μL
Content And Storage Store the kit at -20°C. For best results, store the components as described in the protocol.
For Use With (Application) Quantification
Product Type F2 (Pig) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.