Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ F3F ELISA Kit

Catalog No. 89100364 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-364 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-364 Supplier Abnova™ Supplier No. KA2099

Includes

  • PLT, 96 wells
  • STD, 500μL
  • CTL1, 250μL
  • CTL2, 250μL
  • DIL, 35mL
  • TMB, 15mL
  • STOP, 15mL
  • WASH, 40mL x 2
  • AB1, 150μL
  • AB2, 150μL

Measures a peptide of 11 amino acids (CVCPVSNAMCR) called Fibulin-3 fragment of EFEMP1.

This assay is a competitive immunoassay utilizing a synthetic peptide coated onto the ELISA plate

Specifications

Calibration Range 20 to 500pM
Conjugate HRP
Cross Reactivity Human, Mouse, Rat
Detection Limit 17pM
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Urine
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.