Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ F9 (Human) ELISA Kit

Catalog No. 89100319 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-319 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-319 Supplier Abnova™ Supplier No. KA0472
Add to Cart
Add to Cart

Includes

  • Human Factor IX Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against Factor IX
  • Sealing Tapes
  • Human Factor IX Standard: Human Factor IX in a buffered protein base (lyophilized), 220ng
  • Biotinylated Human Factor IX Antibody (50x)
  • EIA Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

Use to measure quantitatively human F9. Sandwich enzyme immunoassay.

This assay employs a quantitative sandwich enzyme immunoassay technique that measures factor IX in less than 4h

Specifications

Calibration Range 1.56 to 100ng/mL
Conjugate Biotin
Cross Reactivity Human
Description F9 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human F9.
Detection Limit 0.4ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Samples, CSF, Plasma, Serum
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.