Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fabp1 (Mouse) ELISA Kit

Catalog No. 89100662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-662 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-662 Supplier Abnova™ Supplier No. KA2472
Add to Cart
Add to Cart

For quantitative measurement of Fabp1 in serum

Specifications

Conjugate HRP
Cross Reactivity Mouse
Description Fabp1 (Mouse) ELISA Kit is for the quantitative measurement of Fabp1 in serum.
Detection Limit 15.63 to 1000ng/mL
Detection Method Colorimetric
Includes 96 wells Anti-mouse H-FABP antibody coated wells, Enzyme conjugate reagent (11mL), Reference standard stock (1vial), Diluent (25mL), 20X Wash Buffer (50mL), TMB Reagent (11mL), Stop Solution (11mL)
Quantity 1 Kit
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Fabp1 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.