Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FABP3 (Pig) ELISA Kit

Catalog No. 89100667 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-667 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-667 Supplier Abnova™ Supplier No. KA2477
Add to Cart
Add to Cart

Use to measure quantitatively FABP3 in plasma and serum.

FABP3 (Pig) ELISA Kit is an enzyme immunoassay for the quantitative determination of FABP3 in pig serum or plasma.

Quantification

Specifications

Conjugate HRP
Description FABP3 (Pig) ELISA Kit is for the quantitative measurement of FABP3 in plasma and serum.
Quantity 1 Kit
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type FABP3 (Pig) ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.