Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FASLG (Human), Biotin, ELISA Kit

Catalog No. 89100745 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-745 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-745 Supplier Abnova™ Supplier No. KA3038
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Human FASL antibody 96 (8x12) wells
  • Lyophilized FASL standards 10ng x 2
  • Sample / Standard diluent buffer 30mL
  • Biotin conjugated anti-Human FASL antibody (Concentrated), Dilution: 1:100 130 ul
  • antibody diluent buffer 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 130μL
  • ABC diluent buffer 12mL
  • TMB substrate 10mL
  • Stop solution 10mL
  • Wash buffer (25X) 30mL

For quantitative measurement of Human ERBB2

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description FASLG (Human) ELISA Kit is intended for the quantitative measurement of human FASL.
Detection Limit 2 pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body fluid, cell culture supernatant, plasma, serum and tissue lysate
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type FASLG (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.