Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FastLink Avidin Labeling Kit

Catalog No. 89028249 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 10, 3 x 100 μg/KT
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-249 3 x 10, 3 x 100 μg/KT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-249 Supplier Abnova Corporation Supplier No. KA1555

missing translation for 'validMainframeMsg'

FastLink Avidin Labeling Kit is used for simple and rapid preparation of avidin labeled antibodies and proteins

Conjugation

Specifications

For Use With (Application) Conjugation
Includes 96 well FastLink mix, vial modifier reagent, vial quencher reagent
Quantity 3 x 10, 3 x 100 μg/KT
Color Brown
Stability 12 Months
Content And Storage -20°C
Product Type Avidin Labeling Kit
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.