Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FCGR2B (Human) ELISA Kit

Catalog No. 89100457 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-457 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-457 Supplier Abnova™ Supplier No. KA2177

Includes

  • Fcγ RIIB/C Microplate (Item A)
  • Wash Buffer Concentrate (20x) (Item B): 20x concentrated solution
  • Standards (Item C): recombinant human Fcγ RIIB/C
  • Assay Diluent A (Item D)
  • Assay Diluent B (Item E): 5x concentrated buffer. For Standard/Sample (cell culture medium/urine) diluent
  • Detection Antibody Fcγ RIIB/C (Item F): biotinylated antihuman Fcγ RIIB/C (each vial is enough to assay half microplate
  • HRP-Streptavidin Concentrate (Item G): 200x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H)
  • Stop Solution (Item I): 0.2M sulfuric acid

Use to measure quantitatively human FCGR2B. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for human Fcγ RIIB/C coated on a 96-well plate

Specifications

Conjugate HRP
Cross Reactivity Human
Description FCGR2B (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of human FCGR2B.
Detection Limit 85pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 100 μL
Sensitivity <85 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type FCGR2B (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.