Learn More
Specifications
Specifications
| Calibration Range | 0.1 to 5ng/mL |
| Conjugate | HRP |
| Cross Reactivity | Dog |
| Detection Method | Colorimetric |
| Includes | 96-microwell with Polyclonal Anti-Fentanyl, 12mL Fentanyl Conjugate, 2mL Fentanyl positive Refrence standard, 2mL negative standard, 12mL TMB Substrate, 12mL stop reagent |
| Quantity | 96 Tests |
| Sample Type | Oral Fluid, Plasma, Serum, Urine, Whole Blood |
| Sample Volume | 20 μL |
| Content And Storage | 4°C |
| For Use With (Application) | Sensitive in-vitro test to detect the presence of Fentanyl in Canine samples such as whole Blood, Serum, Plasma and Urine |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.