Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fetuin (Human) ELISA Kit

Catalog No. 89100353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-353 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-353 Supplier Abnova™ Supplier No. KA2130
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Diluent Concentrate (Running Buffer): 5X concentrated diluent running buffer
  • Wash Solution Concentrate: 20X concentrated wash solution
  • Enzyme-Antibody Conjugate 100X: affinity purified anti-Human Fetuin antibody conjugated with horseradish peroxidase in a stabilizing buffer
  • Chromogen-Substrate Solution
  • Stop Solution: 0.3M sulfuric acid. WARNING: Avoid contact with skin
  • Anti-Human Fetuin ELISA Micro Plate: Twelve removable eight (8) well micro well strips in well holder frame. Each well is coated with affinity purified anti-Human Fetuin
  • Human Fetuin Calibrator: a lyophilized Human Fetuin calibrator

Measure human Fetuin. Two-site enzyme linked immunoassay.

Specifications

Conjugate HRP
Cross Reactivity Human
Description Fetuin (Human) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of human Fetuin.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.