Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FGF2 (Human) ELISA Kit

Catalog No. 89100201 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-201 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-201 Supplier Abnova™ Supplier No. KA0527
Add to Cart
Add to Cart

Includes

  • bFGF Microtiter plate, 96 wells
  • Biotin Conjugate, 7mL
  • Avidin Conjugate, 14mL
  • bFGF Standard, 2 vials
  • Calibrator diluent I, 25mL
  • Calibrator diluent II, 25mL
  • Wash Buffer (20x), 60mL
  • Substrate A, 10mL
  • Substrate B, 10mL
  • Stop solution, 14mL

Use to measure quantitatively human FGF2. Sandwich enzyme immunoassay.

This bFGF enzyme-linked immunosorbent assay (ELISA) applies a technique called a quantitative sandwich immunoassay

Specifications

Calibration Range 78 to 8000pg/mL
Conjugate HRP
Cross Reactivity Human
Description FGF2 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human FGF2.
Detection Limit 78pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume 100 μL
Sensitivity Calibrator Diluent I: 80 pg/mL; Calibrator Diluent II: 78 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.