Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fgf2 (Mouse) ELISA Kit

Catalog No. 89028512 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-512 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-512 Supplier Abnova™ Supplier No. KA1777

missing translation for 'validMainframeMsg'

Includes

  • bFGF Microplate (Item A): coated with anti-mouse bFGF, 96 wells (12 strips x 8 wells)
  • Wash Buffer Concentrate (Item B): 20X concentrated solution 25mL
  • Standards (Item C): recombinant mouse bFGF 2 vials
  • Assay Diluent A (Item D): diluent buffer, 0.09% sodium azide as preservative. For Standard/Sample (serum/plasma) diluent 30mL
  • Assay Diluent B (Item E): 5X concentrated buffer. For Standard/Sample (cell culture medium) diluent, 15mL
  • Detection Antibody bFGF (Item F): Biotinylated anti-mouse bFGF (each vial is enough to assay half microplate), 2 vials
  • HRP-Streptavidin concentrates (Item G): 300X concentrated, HRP-conjugated Streptavidin. 200μL
  • TMB One-Step Substrate Reagent (Item H): of 3, 3', 5, 5'- tetramethylbenzidine (TMB) in buffered solution 12mL
  • Stop Solution (Item I): 0.2M sulfuric acid 8mL

In-vitro ELISA for the quantitative measurement of Mouse Fgf2

Specifications

Conjugate Biotin
Cross Reactivity Mouse
Description Fgf2 (Mouse) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Fgf2.
Detection Method Colorimetric
Includes 96 Antibody coated microwell, 25mL Wash Buffer 20X,2 standard vials, 30mL assay diluent A, 15mL assay diluent B, 2 detection Antibody MSP vials, 8μL HRP Streptavidian concentrate, 12mL TMB one step Substrate reagent, 8mL stop solution
Quantity 96 Assays
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type Fgf2 (Mouse) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.