Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fibrinogen (Dog) ELISA Kit

Catalog No. 89024521 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-521 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-521 Supplier Abnova™ Supplier No. KA1401
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Dog Fibrinogen in Urine and cell culture media

Specifications

Calibration Range 20 to 200ng/mL
Conjugate Biotin
Cross Reactivity Dog
Description Fibrinogen (Dog) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of dog fibrinogen in urine and cell culture media.
Detection Method Colorimetric
Includes 96 Antibody coated microwell, 800μg Dog FBG standard, Biotinylated canine FBG Antibody vial, 30mL EIA diluent (10X), 2 30mL Wash Buffer (20X), 80μL Streptavidin Peroxidase Conjugate, 8mL Chromogen Substrate, 12mL stop s
Quantity 1 Kit
Sample Type Cell Culture Medium, Urine
Sample Volume 50 μL
Content And Storage Store the kit at 4°C or -20°C.
For Use With (Application) Quantification
Product Type Fibrinogen (Dog) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.