Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fibrinogen (Human) ELISA Kit

Catalog No. 89100245 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Cell Culture Samples, CSF, Milk, Saliva, Urine
Plasma
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-245 Plasma
89-100-244 Cell Culture Samples, CSF, Milk, Saliva, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-245 Supplier Abnova™ Supplier No. KA0475
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Human FBG Microplate: A 96-well polystyrene microplate coated with a murine antibody against FBG
  • Sealing Tapes
  • Human FBG Standard: Human FBG in a buffered protein base (lyophilized), 100μg
  • Biotinylated Human FBG (lyophilized)
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

This assay employs a quantitative competitive enzyme immunoassay technique that measures FBG in less than 3 hours

Specifications

Calibration Range 2.5 to 40ug/mL
Conjugate Biotin
Cross Reactivity Human
Description Fibrinogen (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human fibrinogen in plasma.
Detection Limit 0.10μg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Human FBG Standard (480ug), Biotinylated FBG (1 vial), MIX Diluent Concentrate (10x): 30mL, Wash Buffer Concentrate (20x): 30mL, Streptavidin-Peroxidase Conjugate (SP Conjugate): 80, Chromogen Substrate (8mL), Stop Solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma
Sample Volume 25 μL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.