Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fibrinogen (Mouse) ELISA Kit

Catalog No. 89100231 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Urine
Plasma
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-231 CCS, Urine
89-024-519 Plasma
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-231 Supplier Abnova™ Supplier No. KA0492
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Mouse FBG Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against mouse FBG 1
  • Sealing Tapes: pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay. 3
  • Mouse FBG Standard: Mouse albumin in a buffered protein base, lyophilized) 80μg
  • Biotinylated mouse FBG ( lyophilized) 1 vial
  • MIX Diluent Concentrate (10x): A 10-fold concentrated buffered protein base 30mL
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant 30mL
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate 80μL
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine 8mL
  • Stop Solution: A 0.5 N hydroxychloric acid to stop the chromogen substrate reaction 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Fibrinogen in Plasma

Specifications

Calibration Range 3.13 to 200ng/mL
Cross Reactivity Mouse
Description Fibrinogen (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse fibrinogen.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Fibrinogen (Mouse) ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.