Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fibrinogen (Rat) ELISA Kit

Catalog No. 89100694 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
Sample Volume:
25 μL
50 μL
100 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate Sample Volume
89-100-694 HRP 100 μL
89-024-525 Biotin 25 μL
89-100-219 Biotin 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-694 Supplier Abnova™ Supplier No. KA2504
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Rat Fibrinogen in Plasma

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Fibrinogen (Rat) ELISA Kit is for the quantitative measurement of Fibrinogen in plsma.
Detection Limit 3.9 to 150ng/mL
Detection Method Colorimetric
Includes 96 wells Anti-rat fibrinogen antibody coated wells, Reference standard (2ug/mL); 10X diluent (25mL), Enzyme conjugate reagent (11mL), Diluent (25mL), 20X Wash Buffer (50mL), TMB Reagent (11mL), Stop Solution (11mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Plasma
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Fibrinogen (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.