Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FK-506 ELISA Kit

Catalog No. 89024537 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-537 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-537 Supplier Abnova™ Supplier No. KA1417
Add to Cart
Add to Cart

Competitive binding Enzyme Immunoassay for the quantitative measurement of FK-506

Specifications

Conjugate HRP
Description FK-506 ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of FK-506.
Detection Method Colorimetric
Includes 96-well microtiter plate, 0.9mL negative calibrator, 0.75mL 3 positive calibrator vials, 10.5mL FK-506-HRP Conjugate, 10.5mL Tetramethylbenzidine Substrate, 15mL Wash Buffer, 6mL stop solution
Quantity 1 Kit
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type FK-506 ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.