Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FLT1 (Human) ELISA Kit

Catalog No. 89027024 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma (EDTA), Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-027-024 CCS, Plasma (EDTA), Serum
89-100-824 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-027-024 Supplier Abnova™ Supplier No. KA0665
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human FLT1

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description sFLT1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human soluble FLT1.
Detection Method Colorimetric
Includes 96 wells Antibody coated microwell, 10ng lyophilized recombinant Mouse Fn1 standard vials, 30mL sample diluent Buffer, 130μL Biotinylated Anti-Mouse Fn1 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin Biotin Peroxidase c
Quantity 1 Kit
Sample Type CCS, Plasma (EDTA), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type FLT1 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.