Learn More
Specifications
Specifications
| Calibration Range | 5 to 25ng/mL |
| Conjugate | HRP |
| Description | Flunitrazepam ELISA Kit is a sensitive in-vitro test to detect the presence of Flunitrazepam and 7-amino flunitrazepam in forensic samples such as whole blood, serum, plasma and urine. |
| Detection Method | Colorimetric |
| Includes | 96-microwell coated with Polyclonal anti Flunitrazepam, 7mL positive and negative control 1, 20mL Enzyme Conjugate, 2mL TMB Substrate, 30mL stop solution |
| Quantity | 96 Tests |
| Sample Type | Plasma, Serum, Urine, Whole Blood |
| Sample Volume | 20 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.