Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 350-I Hydrazide

Catalog No. 89139521 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity: 1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-139-521 Quantity: 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-521 Supplier Abnova™ Supplier No. U0235
Add to Cart
Add to Cart

Water soluble fluorescent dye forms stable Schiff Base with carbonyl group

Protein conjugates prepared with Fluorescent Dye 350-I Hydrazide are bright with fluorescence unaffected by pH in the physiological range (pH 4 to 10). Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis

  • Spectral Equivalent: Alexa Fluor 350, AMCA, DyLight 350
  • Purity: ≥90%
  • Molar Extinction Co-efficient: ≥18,000 M-1cm-1
  • Excitation (Max): 345nm
  • Emission (Max): 442nm
  • Solubility: DMSO, water
  • Storage Buffer: DMSO

Application: Conjugation

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent dye that is water-soluble and forms stable Schiff Base with carbonyl group. Protein conjugates prepared with Fluorescent Dye 350-I Hydrazide are bright with fluorescence unaffected by pH in the physiological range (pH 4-10).
For Use With (Application) Conjugation
Quantity 1 mg
Product Type Fluorescent Dye 350-I Hydrazide
Purity ≥90%
Equivalence Alexa Fluor 350, AMCA, DyLight 350
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 350-I Hydrazide
Emission 442
Excitation 345
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.