Learn More
Description
Protein conjugates prepared with Fluorescent Dye 350-I Maleimide are bright with fluorescence unaffected by pH in the physiological range (pH 4 to 10). Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 350, AMCA, DyLight 350
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥18,000 M-1cm-1
- Excitation (Max): 345nm
- Emission (Max): 442nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable thioether bond with free thiol groups. Protein conjugates prepared with Fluorescent Dye 350-I Maleimide are bright with fluorescence unaffected by pH in the physiological range (pH 4-10). |
| For Use With (Application) | Conjugation |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 350-I Maleimide |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 350, AMCA, DyLight 350 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 350-I Maleimide |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.