Learn More
Description
Protein conjugates prepared with Fluorescence Dye 405-I are bright with fluorescence unaffected by pH in the physiological range (pH 4-10). Fluorescence Dye 405-I labeled conjugates are excellent violet laser reagents for flow cytometry research. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Spectral Equivalent: Alexa Fluor 405, DyLight 405
- Molar Extinction Co-efficient: ≥20,000 M-1cm-1
- Excitation (Max): 401nm
- Emission (Max): 420nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 405-I with Phalloidin conjugate. Protein conjugates prepared with Fluorescence Dye 405-I are bright with fluorescence unaffected by pH in the physiological range (pH 4-10). Fluorescence Dye 405-I labeled conjugates are excellent violet laser reagents for flow cytometry research. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye 405-I Phalloidin |
| Sufficient For | 300 Reactions |
| Equivalence | Alexa Fluor 405, DyLight 405 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 405-I Phalloidin |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.