Learn More
Description
Protein conjugates prepared with Fluorescent Dye 405-I Succinimidyl Ester are bright with fluorescence unaffected by pH in the physiological range (pH 4 to 10). Fluorescent Dye 405-I Succinimidyl Ester labeled conjugates are excellent violet laser reagents for flow cytometry research. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 405, DyLight 405
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥20,000 M-1cm-1
- Excitation (Max): 401nm
- Emission (Max): 420nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Protein conjugates prepared with Fluorescent Dye 405-I Succinimidyl Ester are bright with fluorescence unaffected by pH in the physiological range (pH 4-10). Fluorescent Dye 405-I Succinimidyl Ester labeled conjugates are excellent violet laser reagents for flow cytometry research. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 405-I Succinimidyl Ester |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 405, DyLight 405 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 405-I Succinimidyl Ester |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.