Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 450-M Succinimidyl Ester (Violet)

Catalog No. 89139553 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-553 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-553 Supplier Abnova™ Supplier No. U0267
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water-soluble fluorescent dye forms stable peptide bond with protein amino groups

All the Fluorescent Dye M series products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. Besides, in comparison with phycobiliprotein tandems, Fluorescent Dye M series possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.

  • Spectral Equivalent: Pacific Blue
  • Purity: >90%
  • Molar Extinction Co-efficient: >30,000 M-1cm-1
  • Excitation (Max): 403nm
  • Emission (Max): 454nm
  • Recommended Excitation: Violet (405nm)
  • Recommended Filter Sets: 450/50nm Filter
  • Solubility: DMSO, water
  • Storage Buffer: DMSO

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. All the Fluorescent Dye M series products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. Besides, in comparison with phycobiliprotein tandems, Fluorescent Dye M series possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications.
For Use With (Application) Conjugation; Flow Cytometry
Quantity 1 mg
Product Type Fluorescent Dye 450-M Succinimidyl Ester (Violet)
Filter Type 450/50nm
Purity >90%
Equivalence Pacific Blue
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 450-M Succinimidyl Ester (Violet)
Emission 454
Excitation 403
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.