Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 488-I Maleimide

Catalog No. 89139524 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-524 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-524 Supplier Abnova™ Supplier No. U0238
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water soluble fluorescent dye forms stable thioether bond with free thiol groups

Fluorescent Dye 488-I Maleimide conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescent Dye 488-I Maleimide is not affected by pH between 4 to 10. Compared to Alexa Fluor 488, Fluorescent Dye 488-I Maleimide of single isomer has much higher purity, bringing better performance consistency from batch to batch. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis

  • Spectral Equivalent: Alexa Fluor 488, Cy2, DyLight 488, FITC
  • Purity: ≥90%
  • Molar Extinction Co-efficient: >70,000 M-1cm-1
  • Excitation (Max): 491nm
  • Emission (Max): 514nm
  • Solubility: DMSO, water
  • Storage Buffer: DMSO

Application: Conjugation

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent dye that is water-soluble and forms stable thioether bond with free thiol groups. Fluorescent Dye 488-I Maleimide conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescent Dye 488-I Maleimide is not affected by pH between 4-10. Compared to Alexa Fluor 488, Fluorescent Dye 488-I Maleimide of single isomer has much hihger purity, bringing better performance consistancy from batch to batch.
For Use With (Application) Conjugation
Quantity 1 mg
Product Type Fluorescent Dye 488-I Maleimide
Purity ≥90%
Equivalence Alexa Fluor 488, Cy2, DyLight 488, FITC
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 488-I Maleimide
Label Type Other Labels or Dyes
Emission 514
Excitation 491
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.