Learn More
Description
Fluorescent Dye 488-I Succinimidyl Ester conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescent Dye 488-I Succinimidyl Ester is not affected by pH between 4 to 10. Fluorescent Dye 488-I Succinimidyl Ester of single isomer has much higher purity, with better performance consistency from batch to batch. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 488, Cy2, DyLight 488, FITC
- Purity: ≥90%
- Molar Extinction Co-efficient: >70,000 M-1cm-1
- Excitation (Max): 491nm
- Emission (Max): 514nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Fluorescent Dye 488-I Succinimidyl Ester conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescent Dye 488-I Succinimidyl Ester is not affected by pH between 4-10. Compared to Alexa Fluor 488, Fluorescent Dye 488-I Succinimidyl Ester of single isomer has much hihger purity, bringing better performance consistancy from batch to batch. |
| For Use With (Application) | Conjugation |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 488-I Succinimidyl Ester |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 488, Cy2, DyLight 488, FITC |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 488-I Succinimidyl Ester |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.