Learn More
Description
Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Spectral Equivalent: Alexa Fluor 532
- Molar Extinction Co-efficient: ≥80,000 M-1cm-1
- Excitation (Max): 542nm
- Emission (Max): 558nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 532-I with Phalloidin conjugate. |
| For Use With (Application) | Conjugation |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye 532-I Phalloidin |
| Sufficient For | 300 Reactions |
| Equivalence | Alexa Fluor 532 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 532-I Phalloidin |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.