Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 532-I Streptavidin

Catalog No. 89139571 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-571 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-571 Supplier Abnova™ Supplier No. U0285
Add to Cart
Add to Cart

Fluorescence Dye 532-I with Streptavidin conjugate

Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.

  • Spectral Equivalent: Alexa Fluor 532
  • Molar Extinction Co-efficient: ≥80,000 M-1cm-1
  • Excitation (Max): 542nm
  • Emission (Max): 558nm
  • Solubility: Water
  • Storage Buffer: Water

Application: Conjugation

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye 532-I with Streptavidin conjugate.
For Use With (Application) Conjugation
Quantity 200 μg
Product Type Fluorescent Dye 532-I Streptavidin
Equivalence Alexa Fluor 532
Storage Buffer Water
Content And Storage Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C.
Label or Dye Fluorescent Dye 532-I Streptavidin
Emission 558
Excitation 542

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.