Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 532-I Succinimidyl Ester

Catalog No. 89139527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-527 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-527 Supplier Abnova™ Supplier No. U0241
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water-soluble fluorescent dye forms stable peptide bond with protein amino groups

Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis

  • Spectral Equivalent: Alexa Fluor 532
  • Purity: ≥90%
  • Molar Extinction Co-efficient: >80,000 M-1cm-1
  • Excitation (Max): 542nm
  • Emission (Max): 558nm
  • Solubility: DMSO, water
  • Storage Buffer: DMSO

Application: Conjugation

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups.
For Use With (Application) Conjugation
Quantity 1 mg
Product Type Fluorescent Dye 532-I Succinimidyl Ester
Purity ≥90%
Equivalence Alexa Fluor 532
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 532-I Succinimidyl Ester
Label Type Other Labels or Dyes
Emission 558
Excitation 542
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.