Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 550-C Amine

Catalog No. 89139403 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-403 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-403 Supplier Abnova™ Supplier No. U0117

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Fluorescent dye derived from trimethine cyanine with an amino group protected as hydrochloride

  • Bears an intrinsic positive charge and reacts with activated carboxylic acid at pH 7 to 9
  • Chemical Name: 2-((1E,3E)-3-(1-(3-ammoniopropyl)-3,3-dimethylindolin-2-ylidene)prop-1-enyl)-1-ethyl-3,3-dimethyl-3H-indolium chloride iodide
  • Chemical Formula: C28H37CIIN3
  • Mol. Weight: 577.97g/mol
  • Purity: ≥95%
  • Molar Extinction Co-efficient: 1,50,000 M-1cm-1 (CH3OH)
  • Excitation (Max): 548nm (CH3OH)
  • Emission (Max): 567nm (CH3OH)
  • Recommended Excitation: 532nm Nd:YAG, 532nm green diode and 543nm green HeNe
  • Recommended Filter Sets: TRITC
  • Solubility: Soluble in Acetonitrile, DMF, DMSO and Methanol; Slightly soluble in water

Application: Conjugation

Recommended Storage: Store in the dark at -20°C.

Specifications

Description Fluorescent dye derived from trimethine cyanine with an amino group protected as hydrochloride. Bearing an intrinsic positive charge and reacts with activated carboxylic acid at pH 7-9.
Formulation Derived from trimethine cyanine
For Use With (Application) Conjugation
Quantity 5 mg
Product Type Fluorescent Dye 550-C Amine
pH Range 7 to 9
Molecular Weight (g/mol) 577.97g/mol
Light Source 532nm green diode, 543nm green HeNe and 532nm Nd:YAG
Filter Type TRITC
Purity ≥95%
Content And Storage Store in the dark at -20°C
Label or Dye Fluorescent Dye 550-C Amine
Emission 567 (CH3OH)
Excitation 548 (CH3OH)
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.