Learn More
Description
Compared with Cy3 conjugates, those of Fluorescent Dye 555-I Succinimidyl Ester are slightly red-shifted, resulting in an optimal match to filters designed for Cy3 dyes in accompany with improved photostability. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 555, Cy3, DyLight 550, TRITC
- Purity: ≥90%
- Molar Extinction Co-efficient: >1,00,000 M-1cm-1
- Excitation (Max): 555nm
- Emission (Max): 565nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Compared with Cy3 conjugates, those of Fluorescent Dye 555-I Succinimidyl Ester are slightly red-shifted, resulting in an optimal match to filters designed for Cy3 dyes in accompany with improved photostability. Besides, their fluorescence is pH-independent from pH 3 to 11. |
| For Use With (Application) | Conjugation |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 555-I Succinimidyl Ester |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 555, Cy3, DyLight 550, TRITC |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 555-I Succinimidyl Ester |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.