Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 570-N Amine

Catalog No. 89139422 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-422 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-422 Supplier Abnova™ Supplier No. U0136

missing translation for 'validMainframeMsg'

Water insoluble fluorescent dye derived from dipyrrometheneboron difluoride

  • Chemical Formula: C28H30BF2N3O2
  • Mol. Weight: 489.36g/mol
  • Purity: ≥98%
  • Molar Extinction Co-efficient: 50,000 M-1cm-1 (CH3OH)
  • Excitation (Max): 573nm (CH3OH)
  • Emission (Max): 612nm (CH3OH)
  • Recommended Excitation: 568nm Kr1 and 543nm green HeNe
  • Recommended Filter Sets: RITC
  • Solubility: Methanol, DMSO, DMF, acetonitrile, dichloromethane

Application: Conjugation

Recommended Storage: Store in the dark at -20°C.

Specifications

Description Fluorescent dye derived from dipyrrometheneboron difluoride. Water insoluble and forms stable amide bond with activated carboxylic acid at pH 7-9.
Formulation Derived from dipyrrometheneboron difluoride
For Use With (Application) Conjugation
Quantity 5 mg
Product Type Fluorescent Dye 570-N Amine
pH Range 7 to 9
Molecular Weight (g/mol) 489.36g/mol
Light Source 568nm Kr1 and 543nm green HeNe
Filter Type RITC
Purity ≥98%
Content And Storage Store in the dark at -20°C
Label or Dye Fluorescent Dye 570-N Amine
Emission 612 (CH3OH)
Excitation 573 (CH3OH)
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.