Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 577-N PEO Amine

Catalog No. 89139427 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-427 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-427 Supplier Abnova™ Supplier No. U0141

Water soluble fluorescent dye with amine group and protected as hydrochloride

  • Chemical Name: 2-(2-{2-[(9-cyano-8-oxo-8H-acenaphtho[1,2-b]pyrrol-3-yl)amino]ethoxy}ethoxy)ethanaminium chloride
  • Chemical Formula: C21H21CIN4O3
  • Mol. Weight: 412.87g/mol
  • Purity: ≥95%
  • Molar Extinction Co-efficient: 60,000 M-1cm-1 (CH3CH2OH)
  • Excitation (Max): 576nm (CH3CH2OH)
  • Emission (Max): 592nm (CH3CH2OH)
  • Recommended Excitation: 568nm Kr1 and 543nm green HeNe
  • Recommended Filter Sets: RITC
  • Solubility: Methanol, DMSO, DMF, acetonitrile

Application: Conjugation

Recommended Storage: Store in the dark at -20°C.

Specifications

Description Fluorescent dye with amine group and protected as hydrochloride. Water soluble and reacts with activated carboxylic acid at pH 7-9.
For Use With (Application) Conjugation
Quantity 5 mg
Product Type Fluorescent Dye 577-N PEO Amine
pH Range 7 to 9
Molecular Weight (g/mol) 412.87g/mol
Light Source 568nm Kr1 and 543nm green HeNe
Filter Type RITC
Purity ≥95%
Content And Storage Store in the dark at -20°C
Label or Dye Fluorescent Dye 577-N PEO Amine
Emission 592 (CH3CH2OH)
Excitation 576 (CH3CH2OH)
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.