Learn More
Description
Biomolecules conjugated to Fluorescent Dye 594-I Succinimidyl Ester exhibit little spectral overlap with green-fluorescent conjugates, and can be efficiently excited by 568nm line of Ar-Kr laser and by the 594nm line of orange He-Ne laser. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 594, DyLight 594, Texas Red
- Purity: ≥90%
- Molar Extinction Co-efficient: >80,000 M-1cm-1
- Excitation (Max): 592nm
- Emission (Max): 614nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Biomolecules conjugated to Fluorescent Dye 594-I Succinimidyl Ester exhibit little spectral overlap with green-fluorescent conjugates, and can be efficiently excited by 568nm line of Ar-Kr laser and by the 594nm line of orange He-Ne laser. |
| For Use With (Application) | Conjugation |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 594-I Succinimidyl Ester |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 594, DyLight 594, Texas Red |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 594-I Succinimidyl Ester |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.