Learn More
Description
Fluorescence emission of Fluorescence Dye 633-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I and R-phycoerythrin, making it ideal for multi-color labeling. In comparison with Alexa Fluor 633, the extinction coefficient of Fluorescence Dye 633-I is much higher. Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.
- Spectral Equivalent: Alexa Fluor 633, DyLight 633
- Molar Extinction Co-efficient: ≥2,40,000 M-1cm-1
- Excitation (Max): 638nm
- Emission (Max): 655nm
- Solubility: Water
- Storage Buffer: Water
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 633-I with Streptavidin conjugate. Fluorescence emission of Fluorescence Dye 633-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I and R-phycoerythrin, making it ideal for multi-color labeling. In comparison with Alexa Fluor 633, the extinction coefficient of Fluorescence Dye 633-I is much higher (250,000 cm-1M-1). |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 200 μg |
| Product Type | Fluorescent Dye 633-I Streptavidin |
| Equivalence | Alexa Fluor 633, DyLight 633 |
| Storage Buffer | Water |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C. |
| Label or Dye | Fluorescent Dye 633-I Streptavidin |
| Label Type | Other Labels or Dyes |
| Emission | 655 |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.