Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 633-I Succinimidyl Ester

Catalog No. 89139533 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-533 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-533 Supplier Abnova™ Supplier No. U0247
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water-soluble fluorescent dye forms stable peptide bond with protein amino groups

Fluorescence emission of Fluorescent Dye 633-I Succinimidyl Ester is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I and R-phycoerythrin, making it ideal for multi-color labeling. In comparison with Alexa Fluor 633, the extinction coefficient of Fluorescent Dye 633-I Succinimidyl Ester is much higher. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis

  • Spectral Equivalent: Alexa Fluor 633, DyLight 633
  • Purity: ≥90%
  • Molar Extinction Co-efficient: >2,40,000 M-1cm-1
  • Excitation (Max): 638nm
  • Emission (Max): 655nm
  • Solubility: DMSO, water
  • Storage Buffer: DMSO

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Fluorescence emission of Fluorescent Dye 633-I Succinimidyl Ester is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I and R-phycoerythrin, making it ideal for multi-color labeling. In comparison with Alexa Fluor 633, the extinction coefficient of Fluorescent Dye 633-I Succinimidyl Ester is much higher (250,000 cm-1M-1).
For Use With (Application) Conjugation; Flow Cytometry
Quantity 1 mg
Product Type Fluorescent Dye 633-I Succinimidyl Ester
Purity ≥90%
Equivalence Alexa Fluor 633, DyLight 633
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 633-I Succinimidyl Ester
Emission 655
Excitation 638
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.