Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 645-A Amine

Catalog No. 89139447 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-447 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-447 Supplier Abnova™ Supplier No. U0161
Add to Cart
Add to Cart

Water soluble fluorescent dye derived from Cyanine

  • Chemical Name: 2-((1E,3E,5E)-5-(1-(3-ammoniopropyl)-3,3-dimethyl-5-sulfonatoindolin-2-ylidene)penta-1,3-dienyl)-1-ethyl-3,3-dimethyl-3H-indolium-5-sulfonate
  • Chemical Formula: C30H37N3O6S2
  • Mol. Weight: 599.76g/mol
  • Purity: ≥95%
  • Molar Extinction Co-efficient: 2,50,000 M-1cm-1 (CH3COH)
  • Excitation (Max): 643nm (CH3OH)
  • Emission (Max): 662nm (CH3OH)
  • Recommended Excitation: Argon-Krypton ion laser 647nm, Red diode laser 635nm
  • Recommended Filter Sets: Cy5, APC
  • Solubility: Methanol, DMSO, DMF, water, acetonitrile

Application: Conjugation

Recommended Storage: Store in the dark at -20°C.

Specifications

Description Fluorescent dye derived from Cyanine. Water soluble and reacts toward activated carboxylic acid at pH 7-9 inserting a negative charge in the labeled molecule.
Formulation Derived from Cyanine
For Use With (Application) Conjugation
Quantity 5 mg
Product Type Fluorescent Dye 645-A Amine
pH Range 7 to 9
Molecular Weight (g/mol) 599.76g/mol
Light Source Argon-Krypton ion laser 647nm, Red diode laser 635nm
Filter Type APC, Cy5
Purity ≥95%
Content And Storage Store in the dark at -20°C
Label or Dye Fluorescent Dye 645-A Amine
Emission 662 (CH3OH)
Excitation 643 (CH3OH)
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.