Learn More
Description
The slight red shift in absorption spectrum makes Fluorescent Dye 647-I Maleimide an optimal match to filters designed Cy5 dyes. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 647, Cy5, DyLight 650
- Purity: ≥90%
- Molar Extinction Co-efficient: >2,40,000 M-1cm-1
- Excitation (Max): 649nm
- Emission (Max): 664nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable thioether bond with free thiol groups. The slight red shift in absorption spectrum makes Fluorescent Dye 647-I Maleimide an optimal match to filters designed Cy5 dyes.Besides, their fluorescence is pH-independent from pH 3 to 11. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 647-I Maleimide |
| Purity | ≥90% |
| Equivalence | Alexa Fluor 647, Cy5, DyLight 650 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 647-I Maleimide |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.