Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 647-I Streptavidin

Catalog No. 89139583 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-583 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-583 Supplier Abnova™ Supplier No. U0297
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescence Dye 647-I with Streptavidin conjugate

The slight red shift in absorption spectrum makes Fluorescence Dye 647-I an optimal match to filters designed Cy5 dyes. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.

  • Spectral Equivalent: Alexa Fluor 647, Cy5, DyLight 650
  • Molar Extinction Co-efficient: ≥2,40,000 M-1cm-1
  • Excitation (Max): 649nm
  • Emission (Max): 664nm
  • Solubility: Water
  • Storage Buffer: Water

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye 647-I with Streptavidin conjugate. The slight red shift in absorption spectrum makes Fluorescence Dye 647-I an optimal match to filters designed Cy5 dyes.Besides, their fluorescence is pH-independent from pH 3 to 11.
For Use With (Application) Conjugation; Flow Cytometry
Quantity 200 μg
Product Type Fluorescent Dye 647-I Streptavidin
Equivalence Alexa Fluor 647, Cy5, DyLight 650
Storage Buffer Water
Content And Storage Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C.
Label or Dye Fluorescent Dye 647-I Streptavidin
Emission 664
Excitation 649

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.