Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 649-C5 Acid

Catalog No. 89139493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-493 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-493 Supplier Abnova™ Supplier No. U0207
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanine 5 dyes are common red fluorophores, and can tolerate a pH range of 3 to 10

Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis

  • Chemical Formula: C35H41F3N2O10S2
  • Mol. Weight: 770.83g/mol
  • Purity: ≥90%
  • Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
  • Excitation (Max): 649nm
  • Emission (Max): 665nm
  • Solubility: DMSO, water

Application: Conjugation, In vivo imaging

Recommended Storage: Store in the dark at -20°C.

Specifications

Description Cyanine 5 dyes are common red fluorophores, and can tolerate a pH range of 3-10.
For Use With (Application) Conjugation; in vivo Imaging
Quantity 5 mg
Product Type Fluorescent Dye 649-C5 Acid
Molecular Weight (g/mol) 770.83g/mol
Purity ≥90%
Content And Storage Store in the dark at -20°C
Label or Dye Fluorescent Dye 649-C5 Acid
Emission 665
Excitation 649

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.