Learn More
Description
Fluorescence emission of Fluorescent Dye 680-I Hydrazide is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 680, Cy5.5, IRDye 700
- Purity: >90%
- Molar Extinction Co-efficient: >2,00,000 M-1cm-1
- Excitation (Max): 676nm
- Emission (Max): 695nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable Schiff Base with carbonyl group. Fluorescence emission of Fluorescent Dye 680-I Hydrazide is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 680-I Hydrazide |
| Purity | >90% |
| Equivalence | Alexa Fluor 680, Cy5.5, IRDye 700 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 680-I Hydrazide |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.