Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 680-I Phalloidin

Catalog No. 89139586 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
300 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-586 300 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-586 Supplier Abnova™ Supplier No. U0300
Add to Cart
Add to Cart

Fluorescence Dye 680-I with Phalloidin conjugate

Fluorescence emission of Fluorescence Dye 680-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.

  • Spectral Equivalent: Alexa Fluor 680, Cy5.5, IRDye 700
  • Molar Extinction Co-efficient: ≥2,00,000 M-1cm-1
  • Excitation (Max): 676nm
  • Emission (Max): 695nm
  • Solubility: DMSO
  • Storage Buffer: DMSO

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye 680-I with Phalloidin conjugate. Fluorescence emission of Fluorescence Dye 680-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11.
For Use With (Application) Conjugation; Flow Cytometry
Quantity 300 Reactions
Product Type Fluorescent Dye 680-I Phalloidin
Sufficient For 300 Reactions
Equivalence Alexa Fluor 680, Cy5.5, IRDye 700
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 680-I Phalloidin
Emission 695
Excitation 676
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.