Learn More
Description
In comparison with Alexa Fluor 700, Fluorescence Dye 700-I are brighter with stronger absorption at 633nm. Fluorescence emission of Fluorescence Dye 700-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Spectral Equivalent: Alexa Fluor 700
- Molar Extinction Co-efficient: ≥2,00,000 M-1cm-1
- Excitation (Max): 685nm
- Emission (Max): 710nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 700-I with Phalloidin conjugate. In comparison with Alexa Fluor 700, Fluorescence Dye 700-I are brighter with stronger absorption at 633nm. Fluorescence emission of Fluorescence Dye 700-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye 700-I Phalloidin |
| Sufficient For | 300 Reactions |
| Equivalence | Alexa Fluor 700 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 700-I Phalloidin |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.