Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 700-M Streptavidin

Catalog No. 89139589 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-589 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-589 Supplier Abnova™ Supplier No. U0303
Add to Cart
Add to Cart

Fluorescence Dye 700-M with Streptavidin conjugate

All the Fluorescent Dye M series products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. Besides, in comparison with phycobiliprotein tandems, Fluorescent Dye M series possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications. Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.

  • Spectral Equivalent: APC-Alexa Fluor 700 Tandems
  • Recommended Excitation: Red (633nm)
  • Recommended Filter Sets: Cy5.5 Filter
  • Molar Extinction Co-efficient: ≥2,50,000 M-1cm-1
  • Excitation (Max): 657nm
  • Emission (Max): 700nm
  • Solubility: Water
  • Storage Buffer: Water

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye 700-M with Streptavidin conjugate. All the Fluorescent Dye M series products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. Besides, in comparison with phycobiliprotein tandems, Fluorescent Dye M series possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications.
For Use With (Application) Conjugation; Flow Cytometry
Quantity 100 μg
Product Type Fluorescent Dye 700-M Streptavidin
Filter Type Cy5.5 Filter
Equivalence APC-Alexa Fluor 700 Tandems
Storage Buffer Water
Content And Storage Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C.
Label or Dye Fluorescent Dye 700-M Streptavidin
Emission 700
Excitation 657
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.